Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: VN1R5.
Putative pheromone receptor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G-protein coupled receptor GPCR26; hGPCR26; pheromone receptor; V1R-like 5; V1r-like receptor 5; Vomeronasal type-1 receptor 5
Gene Aliases: V1RL5; VN1R5
UniProt ID: (Human) Q7Z5H4
Entrez Gene ID: (Human) 317705
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support