Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSFFGFGQS VEVEILLNDA ESRKRAEHKT EDGKKEKYFL FYDGETVSGK VSLALKNPNK RLEHQGIKIE FIGQIELYYD RGNHHEFVSL VKDLARPGEI TQSQAFDFEF THVEKPYESY TGQNVKLRYF LRATISRRLN DVVKEMDIVV HTLSTYPELN SSIKMEVGIE DCLHIEFEYN KSKYHLKDVI VGKIYFLLVR IKIKHMEIDI IKRETTGTGP NVYHENDTIA KYEIMDGAPV RGESIPIRLF LAGYELTPTM RDINKKFSVR YYLNLVLIDE EERRYFKQQE VVLWRKGDIV RKSMSHQAAI ASQRFEGTTS LGEVRTPSQL SDNNCRQ (1-336 aa encoded by BC009747)
In mammals, there are two paralogues of yeast Vps26, VPS26A and VPS26B. VPS26 is a component of the retromer complex composed of VPS26 (VPS26A or VPS26B), VPS29, VPS35, SNX1 and SNX2. VPS26A and VPS26B subunits define distinct retromer complexes. The retromer complex is important in recycling transmembrane receptors from endosomes to the trans-Golgi network (TGN).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Vacuolar protein sorting-associated protein 26B; Vesicle protein sorting 26B; VPS26 retromer complex component B
Gene Aliases: 1810012I05Rik; 2310075A12Rik; AI848392; Pep8b; VPS26B
UniProt ID: (Human) Q4G0F5, (Mouse) Q8C0E2
Entrez Gene ID: (Human) 112936, (Rat) 300472, (Mouse) 69091
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support