Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: DSQNKTLLK TLAELYTYDK NYGNALEIYL TLRHKDVFQL IHKHNLFSSI KDKIVLLMDF DSEKAVDMLL DNEDKISIKK VVEELEDRPE LQHVYLHKLF KRDHHKGQRY HEKQISLYAE YDRPNLLPFL RDSTHCPLEK ALEICQQRNF VEETVYLLSR MGNSRSALKM IMEELHDVDK AIEFAKEQDD GELWEDLILY SIDKPPFITG LLNNIGTHVD PILLIHRIKE GMEIPNLRDS LVKILQDYNL QILLREGCKK ILVADSLSLL KKMHRTQMKG VLVDEENICE SCLSPILPSD AAKPFSVVVF HCRHMFHKEC LPMPSMNSAA QFCNICSAKN RGPGSAILEM KK (504-854 aa encoded by BC044851)
Vesicle mediated protein sorting plays an important role in segregation of intracellular molecules into distinct organelles. Genetic studies in yeast have identified more than 40 vacuolar protein sorting (VPS) genes involved in vesicle transport to vacuoles. This gene encodes the human ortholog of yeast Vps41 protein which is also conserved in Drosophila, tomato, and Arabidopsis. Expression studies in yeast and human indicate that this protein may be involved in the formation and fusion of transport vesicles from the Golgi. Several transcript variants encoding different isoforms have been described for this gene, however, the full-length nature of not all is known.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mVAM2; S53; vacuolar assembly protein 41; vacuolar protein sorting 41 homolog; Vacuolar protein sorting-associated protein 41 homolog; VAM2 homolog; VPS41 HOPS complex subunit
Gene Aliases: AI317346; HVPS41; hVps41p; HVSP41; mVam2; Vam2; VPS41
UniProt ID: (Human) P49754, (Mouse) Q5KU39
Entrez Gene ID: (Human) 27072, (Mouse) 218035
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support