Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLPTKNRIKR EKIHERCKNI CPPKDTFERT LLHTHDKSRT DLEQVPKIFM KPDFALDDSL TFNSVLPWSH FNTAGGKGNR DAASSKLLQE KLSHYLDIVE VNIAHQISLR SEAFFHAMTS QHELQDYLRK TSQAVKMLRD KIAQIDKVMC EGSLHILRLA LTRNNCVKVY NKLKLMATVH QTQPTVQVLL STSEFVGALD LIATTQEVLQ QELQGIHSFR HLGSQLCELE KLIDKMMIAE FSTYSHSDLN RPLEDDCQVL EECVINKVSQ TEEIDTDVVV KLADQMRMLN FPQWFDLLKD IFSKFTIFLQ RVKATLNIIH SVVLSVLDKN QRTRELEEIS QQKNAAKDNS LDTEVAYLIH EGMFISDAFG EGELTPIAVD TTSQRNASPN SEPCSSDSVS EPECTTDSSS SKEHTSSSAI PGGVDIMVSE DMKLTDSELG KLANNIQELL YSASDICHDR AVKFLMSRAK DGFLEKLNSM EFITLSRLME TFILDTEQIC GRKSTSLLGA LQSQAIKFVN RFHEERKTKL SLLLDNERWK QADVPAEFQD LVDSLSDGKI ALPEKKSGAT EERKPAEVLI VEGQQYAVVG TVLLLIRIIL EYCQCVDNIP SVTTDMLTRL SDLLKYFNSR SCQLVLGAGA LQVVGLKTIT TKNLALSSRC LQLIVHYIPV IRAHFEARLP PKQYSMLRHF GHITKDYHDH IAEISAKLVA IMDSLFDKLL SKYEVKAPVP SACFRNICKQ MTKMHEAIFD HLPEEQTQML FLRINASYKL HLKKQLSHLN VINDGGPQNG LVTADVAFYT GNLQALKGLK DLDLNMAEIW EQKR
This gene encodes for a protein that in yeast forms part of a trimeric vacuolar-protein-sorting complex that is required for retrograde transport of proteins from prevacuoles to the late Golgi compartment. As in yeast, mammalian Vps54 proteins contain a coiled-coil region and dileucine motifs. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hepatocellular carcinoma protein 8; protein phosphatase 1, regulatory subunit 164; Tumor antigen HOM-HCC-8; Tumor antigen SLP-8p; vacuolar protein sorting 54 homolog; Vacuolar protein sorting-associated protein 54; VPS54 GARP complex subunit
Gene Aliases: HCC8; hVps54L; PPP1R164; SLP-8p; VPS54; VPS54L; WR
UniProt ID: (Human) Q9P1Q0
Entrez Gene ID: (Human) 51542
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support