Flagellin is the subunit protein which polymerizes to form the filaments of bacterial flagella. FlaB/D is not essential for polar flagellar synthesis and swimming motility. Homopolymer of FlaB/D is not able to form a functional filament.