Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Viperin Antibodies

          Invitrogen

          Viperin Polyclonal Antibody

          View all (13) Viperin antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Viperin Polyclonal Antibody

          • Antibody Testing Data (2)
          Viperin Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          Viperin Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Viperin Antibody (PA5-42231) in IHC

          Immunohistochemistry analysis of human kidney tissue using an anti-Viperin polyclonal antibody (Product # PA5-42231). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Viperin Antibody in Immunohistochemistry (IHC)
          Viperin Antibody in Western Blot (WB)

          Product Details

          PA5-42231

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          5 µg/mL
          -

          Immunohistochemistry (IHC)

          5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide directed towards the C-terminal of human RSAD2 (aa 301-350).
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS with 2% sucrose

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2606329

          Product Specific Information

          Peptide sequence: YLILDEYMRF LNCRKGRKDP SKSILDVGVE EAIKFSGFDE KMFLKRGGKY

          Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%

          Target Information

          Viperin is a 43kDa protein which is one of several hundred interferon inducible viral proteins. Elevated expression of viperin is induced by LPS and poly I:C through TLR dependent signaling pathways. Sendai virus mediated viperin expression is induced independently of TLR signaling pathways through RLRs - RIG-I and MAVs (Severa, 2006). Viperin has been shown to play an important role in interferon mediated anti-viral activity in a Hepatitis C virus model system (Helbig, 2011). Viperin is induced in lymphoid cells and dendritic cells (DCs) and highly induced in neutrophils and macrophages during acute LCMV infection in mice (Hinson, ). Viperin functions as a mediator of viral resistance and as a marker for interferon responsiveness making its detection and mechanism of action through specific antibody probes valuable for further studies.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Cytomegalovirus-induced gene 5 protein; Radical S-adenosyl methionine domain-containing protein 2; RSAD2; S-adenosylmethionine-dependent nucleotide dehydratase RSAD2; SAND; Viperin; Virus inhibitory protein, endoplasmic reticulum-associated, interferon-inducible

          View more View less

          Gene Aliases: 2510004L01Rik; cig33; CIG5; RSAD2; vig1

          View more View less

          UniProt ID: (Human) Q8WXG1

          View more View less

          Entrez Gene ID: (Human) 91543

          View more View less

          Function(s)
          catalytic activity protein binding protein self-association metal ion binding 4 iron, 4 sulfur cluster binding
          Process(es)
          response to virus positive regulation of toll-like receptor 7 signaling pathway positive regulation of toll-like receptor 9 signaling pathway CD4-positive, alpha-beta T cell activation CD4-positive, alpha-beta T cell differentiation negative regulation of viral genome replication negative regulation of protein secretion defense response to virus type I interferon signaling pathway positive regulation of T-helper 2 cell cytokine production
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-k2m5x:80/100.66.128.182:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0