Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 6
Positive control: Human prostate tissue, human fallopian tube tissue, mouse brain tissue, HepG2 cell lysate, 293 cell lysate, K562 cell lysate, Hela, HepG2.
Predicted band size: 37 kDa
Subcellular Location: Nucleus, Cytoplasm.
WDR77 is a component of the 20S PRMT5-containing methyltransferase complex, which modifies specific arginines to dimethylarginines in several spliceosomal Sm proteins. This modification targets Sm proteins to the survival of motor neurons (SMN) complex for assembly into small nuclear ribonucleoprotein core particles.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Androgen receptor cofactor p44; MEP-50; Methylosome protein 50; Methylosome protein WDR77; MGC2722; OTTHUMP00000013473; p44/Mep50; RP11-552M11.3; testis tissue sperm-binding protein Li 44a; WD repeat-containing protein 77
Gene Aliases: HKMT1069; MEP-50; MEP50; Nbla10071; p44; p44/Mep50; WD45; WDR77
UniProt ID: (Human) Q9BQA1
Entrez Gene ID: (Human) 79084
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support