Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MEPHLLGLLL GLLLGGTRVL AGYPIWWSLA LGQQYTSLGS QPLLCGSIPG LVPKQLRFCR NYIEIMPSVA EGVKLGIQEC QHQFRGRRWN CTTIDDSLAI FGPVLDKATR ESAFVHAIAS AGVAFAVTRS CAEGTSTICG CDSHHKGPPG EGWKWGGCSE DADFGVLVSR EFADARENRP DARSAMNKHN NEAGRTTILD HMHLKCKCHG LSGSCEVKTC WWAQPDFRAI GDFLKDKYDS ASEMVVEKHR ESRGWVETLR AKYSLFKPPT ERDLVYYENS PNFCEPNPET GSFGTRDRTC NVTSHGIDGC DLLCCGRGHN TRTEKRKEKC HCIFHWCCYV SCQECIRIYD VHTCK
WNT3, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC131950; MGC138321; MGC138323; Proto-oncogene Int-4 homolog; Proto-oncogene Wnt-3; wingless-type MMTV integration site family member 3; wingless-type MMTV integration site family, member 3; WNT-3 proto-oncogene protein
Gene Aliases: INT4; TETAMS; WNT3
UniProt ID: (Human) P56703
Entrez Gene ID: (Human) 7473
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support