Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: GEGSQMGLDE CQFQFRNGRW NCSALGERTV FGKELKVGSR EAAFTYAIIA AGVAHAITAA CTQGNLSDCG CDKEKQGQYH RDEGWKWG; Positive Samples: Mouse kidney, Rat kidney; Cellular Location: extracellular matrix, extracellular space
Wnt-7a belongs to the Wnt family of signaling proteins that play a key role in maintaining the integrity of embryonic and adult tissues. It is expressed in placenta, kidney, testis, uterus, fetal lung, and fetal and adult brain. Most Wnt proteins can signal though a mechanism called the canonical Wnt pathway, in which Wnt proteins bind to and activate seven-pass transmembrane receptors of the Frizzled family, ultimately leading to the disruption of ß-catenin degradation. Intracellular accumulation of ß-catenin increases translocation of the protein into the nucleus, where it binds to TCF/LEF transcription factors to induce the expression of numerous genes. Increased Wnt/ß-catenin signaling is associated with tumorigenesis in a diverse set of human cancers. However, Wnt-7a/Frizzled-9 signaling has been shown to act as a tumor suppressor in non-small cell lung cancers.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: postaxial hemimelia; Protein Wnt-7a; wingless-related MMTV integration site 7A; wingless-type MMTV integration site 7A
Gene Aliases: AI849442; px; tw; Wnt-7a; Wnt7a
UniProt ID: (Mouse) P24383
Entrez Gene ID: (Rat) 114850, (Mouse) 22421
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support