Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
In direct ELISAs, no cross-reactivity with recombinant human Wnt-2, -3a, -6, -7a, -7b, recombinant mouse Wnt-1, -2b, -4, -5a, -5b, -8a, -8b, -9b, -10a, -10b, -11, or -16 is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
WNT9A is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. WNT9A is likely to signal over only few cell diameters.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC138165; MGC141991; Protein Wnt-14; Protein Wnt-9a; wingless-type MMTV integration site family member 9A; wingless-type MMTV integration site family, member 14; wingless-type MMTV integration site family, member 9A
Gene Aliases: WNT14; WNT9A
UniProt ID: (Human) O14904
Entrez Gene ID: (Human) 7483
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support