Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • XPNPEP3 Antibodies

          OriGene

          XPNPEP3 Monoclonal Antibody (OTI2D5), TrueMAB™

          View all (13) XPNPEP3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite XPNPEP3 Monoclonal Antibody (OTI2D5), TrueMAB™

          • Antibody Testing Data (6)
          XPNPEP3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          XPNPEP3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          XPNPEP3 Antibody (TA503349) in ICC/IF

          Anti-XPNPEP3 mouse monoclonal antibody (TA503349) immunofluorescent staining of COS7 cells transiently transfected by pCMV6-ENTRY XPNPEP3(RC200888). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          XPNPEP3 Antibody in Immunocytochemistry (ICC/IF)
          XPNPEP3 Antibody in Immunocytochemistry (ICC/IF)
          XPNPEP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          XPNPEP3 Antibody in Western Blot (WB)
          XPNPEP3 Antibody in Western Blot (WB)
          XPNPEP3 Antibody in Flow Cytometry (Flow)

          Product Details

          TA503349

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:150
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:100
          -

          Flow Cytometry (Flow)

          1:100
          -
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          OTI2D5

          Immunogen

          Full length human recombinant protein of human XPNPEP3 produced in HEK293 cell.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.81 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS with 1% BSA, 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          XPNPEP3, also known as X-Prolyl Aminopeptidase 3, is a gene located on chromosome 22q that encodes an enzyme involved in the processing of mitochondrial matrix proteins. The enzyme is classified under the M24B subfamily of metallopeptidases, which specifically remove the N-terminal amino acid from oligopeptides where the penultimate residue is proline. XPNPEP3 is implicated in conditions such as cystic kidney disease and participates in TNF-TNFR2 cellular signaling pathways. Its yeast and plant homologs, like Icp55, perform similar functions by stabilizing proteins after import into the mitochondrial matrix. Mutations in XPNPEP3 are associated with defects in ciliary structure and function, contributing to the phenotypic spectrum seen in nephronophthisis-related ciliopathies (NPHP-RCs). The crystal structure of XPNPEP3 has been resolved at 1.65 A resolution, revealing its interaction with inhibitors such as apstatin. Furthermore, XPNPEP3 is identified as a transcriptional target of the canonical Wnt/beta-catenin signaling pathway, with elevated transcript levels observed in tumors exhibiting activated Wnt signaling.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Aminopeptidase P3; APP3; Intermediate Cleaving Peptidase 55; probable Xaa-Pro aminopeptidase 3; X-Pro aminopeptidase 3; X-prolyl aminopeptidase (aminopeptidase P) 3, putative; X-prolyl aminopeptidase 3, mitochondrial; Xaa-Pro aminopeptidase 3

          View more View less

          Gene Aliases: APP3; E430012M05Rik; ICP55; NPHPL1; XPNPEP3

          View more View less

          UniProt ID: (Human) Q9NQH7, (Rat) B5DEQ3, (Mouse) B7ZMP1

          View more View less

          Entrez Gene ID: (Dog) 481237, (Human) 63929, (Rat) 685823, (Mouse) 321003

          View more View less

          Function(s)
          aminopeptidase activity metallopeptidase activity manganese ion binding hydrolase activity metal ion binding
          Process(es)
          glomerular filtration proteolysis protein processing
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0