Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: SLRLEVKLGQ GCFGEVWMGT WNGTTKVAIK TLKPGTMMPE AFLQEAQIMK KLRHDKLVPL YAVVSEEPIY IVTEFMSKGS LLDFLKEGDG KYLKLPQLVD MAAQIADGMA YIERMNYIHR DLRAANILVG ENLVCKIADF GLARLIEDNE YTARQGAKFP IKWTAPEAAL YGRFTIKSDV WSFGILQTEL VTKGRVPYPG MVNREVLEQV ERGYRMPCPQ GCPESLHELM NLCWKKDPDE RPTFEYIQSF LEDY
YES1 is a recombinant human protein kinase that is a member of the SRC family of non-receptor tyrosine kinases and is the cellular homolog of the Yamaguchi sarcoma virus oncogene. It is involved in the regulation of cell growth and survival, including roles in apoptosis, cell-cell adhesion, cytoskeleton remodeling, and differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-Src; cellular yes-1 protein; EC 2.7.10.2; p60-Src; p61-Yes; pp60c-src; Proto-oncogene c-Yes; proto-oncogene tyrosine-protein kinase YES; Tyrosine-protein kinase Yes; v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1; Yamagichi sarcoma viral (v-yes-1) oncogene homolog 1; Yamaguchi sarcoma oncogene; Yamaguchi sarcoma viral oncogene homolog 1; YES1 proto-oncogene, Src family tyrosine kinase
Gene Aliases: c-Yes; HsT441; p60c-yes; P61-YES; YES; YES1
UniProt ID: (Human) P07947
Entrez Gene ID: (Human) 7525, (Rat) 24884
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support