Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEFVPGGSIS SIINRFGPLP EMVFCKYTKQ ILQGVAYLHE NCVVHRDIKG NNVMLMPTGI IKLIDFGCAR RLAWAGLNGT HSDMLKSMHG TPYWMAPEVI NESGYGRKSD IWSIGCTVFE MATGKPPLAS MDRMAAMFYI GAHRGLMPPL PDHFSENAAD FVRMCLTR
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitogen-activated protein kinase kinase kinase 19; regulated in COPD kinase; Regulated in COPD, protein kinase; SPS1/STE20-related protein kinase YSK4; yeast Sps1/Ste20-related kinase 4; YSK4 Sps1/Ste20-related kinase homolog
Gene Aliases: MAP3K19; RCK; YSK4
UniProt ID: (Human) Q56UN5
Entrez Gene ID: (Human) 80122
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support