Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MENKSLESSQ TDLKLVAHPR AKSKVWKYFG FDTNAEGCIL QWKKIYCRIC MAQIAYSGNT SNLSYHLEKN HPEEFCEFVK SNTEQMREAF ATAFSKLKPE SSQQPGQDAL AVKAGHGYDS KKQQELTAAV LGLICEGLYP ASIVDEPTFK VLLKTADPRY ELPSRKYIST KAIPEKYGAV REVILKELAE ATWCGISTDM WRSENQNRAY VTLAAHFLGL GAPNCLSMGS RCLKTFEVPE ENTAETITRV LYEVFIEWGI SAKVFGATTN YGKDIVKACS LLDVAVHMPC LGHTFNAGIQ QAFQLPKLGA LLSRCRKLVE YFQQSAVAMY MLYEKQKQQN VAHCMLVSNR VSWWGSTLAM LQRLKEQQFV IAGVLVEDSN NHHLMLEASE WATIEGLVEL LQPFKQVAEM LSASRYPTIS MVKPLLHMLL NTTLNIKETD SKELSMAKEV IAKELSKTYQ ETPEIDMFLN VATFLDPRYK RLPFLSAFER QQVENRVVEE AKGLLDKVKD GGYRPAEDKI FPVPEEPPVK KLMRTSTPPP ASVINNMLAE IFCQTGGVED QEEWHAQVVE ELSNFKSQKV LGLNEDPLKW WSDRLALFPL LPKVLQKYWC VTATRVAPER LFGSAANVVS AKRNRLAPAH VDEQVFLYEN ARSGAEAEPE DQDEGEWGLD QEQVFSLGDG VSGGFFGIRD SSFL
ZBED1, also known as DREF, is the human homolog of the Drosophila protein DREF, a transcriptional regulatory factor required for expression of genes involved in DNA replication and cell proliferation. RNA interference (RNAi) experiments targeting ZBED1 RNA resulted in the inhibition of S phase entry and reduction of histone H1 mRNA in HeLa cells, suggesting that ZBED1 also plays a role in cell proliferation in human cells. At least 22 ribosomal proteins (RPs) possess ZBED1 binding sites in their promoters; anti-ZBED1 RNAi also resulted in decreased RP gene expression. Immunoprecipitation analyses have shown that ZBED1 self-associates in vivo via a carboxyl-terminal hATC domain, and this self-association is required for nuclear localization and DNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ac-like transposable element; BED-type zinc finger domain-containing protein 1; DNA replication-related element binding factor; DNA replication-related element-binding factor; dREF homolog; E3 SUMO-protein ligase ZBED1; Putative Ac-like transposable element; Zinc finger BED domain-containing protein 1; zinc finger, BED-type containing 1
Gene Aliases: ALTE; DREF; hDREF; KIAA0785; TRAMP; ZBED1
UniProt ID: (Human) O96006
Entrez Gene ID: (Human) 9189
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support