Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
This antibody is predicted to react in Mouse.
The function of Anti-ZBTB7C has not yet been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Affected by papillomavirus DNA integration in ME180 cells protein 1; APM-1; B230208J24Rik; zinc finger and BTB domain containing 36; zinc finger and BTB domain-containing 7C; Zinc finger and BTB domain-containing protein 36; Zinc finger and BTB domain-containing protein 7C; Zinc finger protein 857C
Gene Aliases: APM-1; APM1; ZBTB36; ZBTB7C; ZNF857C
UniProt ID: (Human) A1YPR0
Entrez Gene ID: (Human) 201501
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support