Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: NSFSMSESKR DLSTSTSREG TPLNNSNSSL LLMNGPGSLF ASENFLGISS
Sequence homology: Cow: 93%; Dog: 79%; Guinea Pig: 93%; Horse: 86%; Human: 100%; Mouse: 92%; Rabbit: 93%; Rat: 93%
ZC3H7A contains 1 C2H2-type zinc finger, 3 C3H1-type zinc fingers and 3 TPR repeats. The function of ZC3H7A remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20318; Zinc finger CCCH domain-containing protein 7A; zinc finger CCCH type domain containing 7; zinc finger CCCH-type domain containing 7; zinc-finger protein AY163807
Gene Aliases: HSPC055; ZC3H7; ZC3H7A; ZC3HDC7
UniProt ID: (Human) Q8IWR0
Entrez Gene ID: (Human) 29066
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support