Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEVVLPLDPA VPAPLCPHGP TLLFVKVTQG KEETRRFYAC SACRDRKDCN FFQWEDEKLS GARLAAREAH NRRCQPPLSR TQCVERYLKF IELPLTQRKF CQTCQQLLLP DDWGQHSEHQ VLGNVSITQL RRPSQLLYPL ENKKTNAQYL FADRSCQFLV DLLSALGFRR VLCVGTPRLH ELIKLTASGD KKSNIKSLLL DIDFR
The ZCCHC4 gene encodes zinc finger CCHC domain-containing protein 4, a member of the zinc finger CCHC-type (ZCCHC) superfamily characterized by a consensus sequence C-X2-C-X4-H-X4-C. This gene plays a vital role in binding single-stranded nucleic acids, particularly RNAs, and is involved in multiple cellular processes. Notably, ZCCHC4 has been implicated in cancer, with aberrantly high expression linked to poor prognosis and chemoresistance in various cancers, including hepatocellular carcinoma, pancreatic cancer, colon cancer, and esophageal cancer. The protein exhibits RNA-modifying capabilities, influencing cell growth and survival. Recent studies have identified the structural mechanisms through which ZCCHC4 interacts with RNA substrates, offering insights into its role in regulating cancer progression and therapeutic resistance. In esophageal cancer, downregulation of ZCCHC4 may enhance sensitivity to chemotherapeutic agents like cisplatin, indicating its potential as a target for cancer treatment strategies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DHHC domain-containing zinc finger protein; rRNA N(6)-adenosine-methyltransferase ZCCHC4; Zinc finger CCHC domain-containing protein 4; zinc finger, CCHC domain containing 4; zinc finger, GRF-type containing 4
Gene Aliases: HSPC052; ZCCHC4; ZGRF4
UniProt ID: (Human) Q9H5U6
Entrez Gene ID: (Human) 29063
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support