Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTQKGSMKPV KKKKTEEPEL EPLCCCEYID RNGEKNHVAT CLCDCQDLDE GCDRWITCKS LQPETCERIM DTISDRLRIP WLRGAKKVNI SIIPPLVLLP VFLHVASWHF LLGVVVLTSL PVLALWYYYL THRRKEQTLF FLSLGLFSLG YMYYVFLQEV VPKGRVGPVQ LAVLTCGLFL ILLALHRAKK NPGYLSNPAS GDRSLSSSQL ECLSRKGQEK TKGFPGADMS GSLNNRTTKD DPKGSSKMPA GSPTKAKEDW CAKCQLVRPA RAWHCRICGI CVRRMDHHCV WINSCVGESN HQAFILALLI FLLTSVYGIT LTLDTICRDR SVFTALFYCP GVYANYSSAL SFTCVWYSVI ITAGMAYIFL IQLINISYNV TEREVQQALR QKTGRRLLCG LIVDTGLLG
Palmitoyltransferase that mediates palmitoylation of KCNMA1, regulating localization of KCNMA1 to the plasma membrane. May be involved in NOS1 regulation and targeting to the synaptic membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DHHC-23; Palmitoyltransferase ZDHHC23; Zinc finger DHHC domain-containing protein 23
Gene Aliases: DHHC-23; NIDD; ZDHHC23
UniProt ID: (Human) Q8IYP9
Entrez Gene ID: (Human) 254887
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support