Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive test controls include: HepG2, Mouse liver, Mouse brain, Rat liver.
Immunogen sequence: MSRRKQSNPR QIKRSLGDME AREEVQLVGA SHMEQKATAP EAPSPPSADV NSPPPLPPPT SPGGPKELEG QEPEPRPTEE EPGSPWSGPD ELEPVVQDGQ RRIRARLSLA TGLSWGPFHG SVQTRASSPR QAEPSPALTL LLVDEACWLR TLPQALTEAE ANTEIHRKDD ALWCRVTKPV PAGGLLSVLL TAEPHSTPGH PVKKEPAEPT CPAPAHDLQL LPQQAGMASI
ZFPM1 is a transcription regulator that plays an essential role in erythroid and megakaryocytic cell differentiation. ZFPM1 is an essential cofactor that acts via the formation of a heterodimer with transcription factors of the GATA family GATA1, GATA2 and GATA3. Such heterodimer can both activate or repress transcriptional activity, depending on the cell and promoter context. The heterodimer formed with GATA proteins is essential to activate expression of genes such as NFE2, ITGA2B, alpha- and beta-globin, while it represses expression of KLF1. ZFPM1 may be involved in regulation of some genes in gonads. ZFPM1 may also be involved in cardiac development, in a non-redundant way with ZFPM2/FOG2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FOG-1; Friend of GATA 1; Friend of GATA protein 1; Friend of GATA-1; Zinc finger protein 89A; Zinc finger protein multitype 1; Zinc finger protein ZFPM1; zinc finger protein, multitype 1
Gene Aliases: FOG; FOG1; ZC2HC11A; ZFN89A; ZFPM1; ZNF408; ZNF89A
UniProt ID: (Human) Q8IX07, (Mouse) O35615
Entrez Gene ID: (Human) 161882, (Rat) 691504, (Mouse) 22761
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support