Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • ZHX2 Antibodies

          Invitrogen

          ZHX2 Recombinant Rabbit Monoclonal Antibody (8N7O2)

          View all (26) ZHX2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ZHX2 Recombinant Rabbit Monoclonal Antibody (8N7O2)

          • Antibody Testing Data (1)
          ZHX2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ZHX2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ZHX2 Antibody (MA5-55598) in WB

          Western Blot analysis of various lysates using ZHX2 Recombinant Monoclonal Antibody (Product # MA5-55598) at 1:1,000 dilution. Secondary antibody: HRP-conjugated Goat anti-Rabbit IgG (H+L) at 1:10,000 dilution. Lysates/proteins: 25 μg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Enhanced Kit. Exposure time: 45s. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ZHX2 Antibody in Western Blot (WB)

          Product Details

          MA5-55598

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          8N7O2

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 260-500 of human ZHX2.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.7 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.05% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          RRID

          AB_3667837

          Product Specific Information

          Immunogen Sequence: VPLNTTKYNS ALDTNATMIN SFNKFPYPTQ AELSWLTAAS KHPEEHIRIW FATQRLKHGI SWSPEEVEEA RKKMFNGTIQ SVPPTITVLP AQLAPTKVTQ PILQTALPCQ ILGQTSLVLT QVTSGSTTVS CSPITLAVAG VTNHGQKRPL VTPQAAPEPK RPHIAQVPEP PPKVANPPLT PASDRKKTKE QIAHLKASFL QSQFPDDAEV YRLIEVTGLA RSEIKKWFSD HRYRCQRGIV H

          Target Information

          Zinc-fingers and homeobox (ZHX) proteins are transcription factors that interact with the activation domain of the A subunit of nuclear factor-Y (NF-YA). ZHX1-3 are ubiquitously expressed proteins expressed in various tissues. They act as transcriptional repressors and localize to the nucleus. The ZHX proteins contain two Cys(2)-His(2)-type zinc-finger motifs and five homeodomains (HDs). These domains allow the ZHX proteins to form homodimers, but they can also form heterodimers with each other. However, this dimerization is not required for repressor activity. Hypermethylation-mediated silencing of ZHX2 is an epigenetic event involved in hepatocellular carcinoma (HCC).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: AFP regulator 1; alpha fetoprotein regulation 1, adult; Alpha-fetoprotein regulator 1; Regulator of AFP; transcription factor ZHX2; Zinc finger and homeodomain protein 2; Zinc fingers and homeoboxes protein 2; zinc-fingers and homeoboxes 2

          View more View less

          Gene Aliases: Afr-1; AFR1; KIAA0854; mKIAA0854; RAF; ZHX2

          View more View less

          UniProt ID: (Human) Q9Y6X8, (Mouse) Q8C0C0

          View more View less

          Entrez Gene ID: (Human) 22882, (Mouse) 387609

          View more View less

          Function(s)
          DNA binding transcription factor activity, sequence-specific DNA binding transcription corepressor activity protein binding identical protein binding protein homodimerization activity metal ion binding protein heterodimerization activity
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter transcription, DNA-templated mRNA catabolic process somatic stem cell population maintenance negative regulation of neuron differentiation negative regulation of transcription, DNA-templated regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter negative regulation of nucleic acid-templated transcription
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0