Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • SLC39A4 Antibodies

          Proteintech

          ZIP4 Polyclonal Antibody

          30 References
          View all (9) SLC39A4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite ZIP4 Polyclonal Antibody

          • Antibody Testing Data (9)
          ZIP4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          ZIP4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 9

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ZIP4 Antibody (20625-1-AP) in IHC (P)

          Immunohistochemistry of paraffin-embedded human liver cancer tissue slide using 20625-1-AP ( ZIP4 antibody at dilution of 1:200 (under 40x lens). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ZIP4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZIP4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZIP4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZIP4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZIP4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ZIP4 Antibody in Western Blot (WB)
          ZIP4 Antibody in Western Blot (WB)
          ZIP4 Antibody in Western Blot (WB)
          ZIP4 Antibody in Western Blot (WB)

          Product Details

          20625-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 14 publications 14 publications

          Immunohistochemistry (IHC)

          -
          View 12 publications 12 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          -
          View 3 publications 3 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Hamster, Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ZIP4 Fusion Protein Ag14524 (23-327 aa encoded by BC062625)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.51 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: SPPAGLLSL LTSGQGALDQ EALGGLLNTL ADRVHCTNGP CGKCLSVEDA LGLGEPEGSG LPPGPVLEAR YVARLSAAAV LYLSNPEGTC EDTRAGLWAS HADHLLALLE SPKALTPGLS WLLQRMQARA AGQTPKTACV DIPQLLEEAV GAGAPGSAGG VLAALLDHVR SGSCFHALPS PQYFVDFVFQ QHSSEVPMTL AELSALMQRL GVGREAHSDH SHRHRGASSR DPVPLISSSN SSSVWDTVCL SARDVMAAYG LSEQAGVTPE AWAQLSPALL QQQLSGACTS QSRPPVQDQL SQSERY (23-327 aa encoded by BC062625)

          Target Information

          The zinc transporter ZIP4, also known as SLC39A4, is a member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP4 was initially identified as zinc-uptake transporter in the intestine and as a candidate gene for acrodermatitis enteropathica (AE), a rare inherited defect in the absorption of dietary zinc. Similar to other ZIP family members, aberrant expression of ZIP4 has been linked to cancer, specifically the progression and aggressiveness of pancreatic cancer. ZIP4 expression has also been shown to be activated in hepatocellular carcinomas, acting to repress apoptosis, enhance cell cycle and increase migration, suggesting ZIP4 may play a role in multiple types of cancers.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Activated in W/Wv mouse stomach 2; mAWMS2; solute carrier family 39 (zinc transporter), member 4; Solute carrier family 39 member 4; Zinc transporter ZIP4; ZIP-4; Zrt- and Irt-like protein 4

          View more View less

          Gene Aliases: 1600025H15Rik; AEZ; AU041686; AWMS2; SLC39A4; ZIP4

          View more View less

          UniProt ID: (Human) Q6P5W5, (Mouse) Q78IQ7

          View more View less

          Entrez Gene ID: (Human) 55630, (Mouse) 72027

          View more View less

          Function(s)
          zinc ion transmembrane transporter activity metal ion transmembrane transporter activity
          Process(es)
          cellular zinc ion homeostasis cellular response to zinc ion starvation zinc II ion transmembrane import transport ion transport zinc II ion transport metal ion transport transmembrane transport zinc II ion transmembrane transport
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-2w6sl:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0