Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: AYITHQRVH TGEKPYECND CGKAFNVNAK LIQHQRIHTG EKPYECNECG KGFRCSSQLR QHQSIHTGEK PYQCKECGKG FNNNTKLIQH QRIHTGEKPY ECTECGKAFS VKGKLIQHQR IHTGEKPYEC NECGKAFRCN SQFRQHLRIH TGEKPYECNE CGKAFSVNGK LMRHQRIHTG EKPFECNECG RCFTSKRNLL DHHRIHTGEK PYQCKECGKA FSINAKLTRH QRIHTGEKPF KCMECEKAFS CSSNYIVHQR IHTGEKPFQC KECGKAFHVN AHLIRHQRSH TGEKPFRCVE CGKGFSFSSD YIIHQTVHTW KKPYMCSVCG KAFRFSFQLS QHQSVHSEGK S (294-643 aa encoded by BC007256)
ZNF23 May be involved in transcriptional regulation. May have a role in embryonic development. Belongs to the krueppel C2H2-type zinc-finger protein family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kruppel-like zinc finger factor X31; Zinc finger protein 23; zinc finger protein 23 (KOX 16); zinc finger protein 32; Zinc finger protein 359; Zinc finger protein 612; Zinc finger protein KOX16
Gene Aliases: B230354B21Rik; KOX16; Zfp23; Zfp612; ZNF23; ZNF359; ZNF612
UniProt ID: (Human) P17027
Entrez Gene ID: (Human) 7571, (Rat) 307839, (Mouse) 234725
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support