Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MSAQSVEEDS ILIIPTPDEE EKILRVKLEE DPDGEEGSSI PWNHLPDPEI FRQRFRQFGY QDSPGPREAV SQLRELCRLW LRPETHTKEQ ILELVVLEQF VAILPKELQT WVRDHHPENG EEAVTVLEDL ESELDDPGQP VSLRRRKREV LVEDMVSQEE AQGLPSSELD AVENQLKWAS WELHSLRHCD DDGRTENGAL APKQELPSAL ESHEVPGTLS MGVPQIFKYG ETCFPKGRFE RKRNPSRKKQ HICDECGKHF SQGSALILHQ RIHSGEKPYG CVECGKAFSR SSILVQHQRV HTGEKPYKCL ECGKAFSQNS GLINHQRIHT GEKPYECVQC GKSYSQSSNL FRHQRRHNAE KLLNVVKV
Transcription factor required for myelination of differentiated oligodendrocytes. Required for the conversion of oligodendrocytes from the premyelinating to the myelinating state. In the developing central nervous system (CNS), involved in the maintenance in the progenitor stage by promoting the cell cycle. Specifically binds to the 5'-TCAT-3' DNA sequence. Has transcription repressor activity in vitro.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Retinoic acid suppression protein A; RSG-A; Zinc finger and SCAN domain-containing protein 3; Zinc finger protein 191; Zinc finger protein 24; zinc finger protein 24 (KOX 17); Zinc finger protein KOX17
Gene Aliases: KOX17; RSG-A; Zfp191; ZNF191; ZNF24; ZSCAN3
UniProt ID: (Human) P17028
Entrez Gene ID: (Human) 7572
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support