Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MQEDRDGSCS TVGGVGYGDS KDCILEPLSL PESPGGTTTL EGSPSVPCIF CEEHFPVAEQ DKLLKHMIIE HKIVIADVKL VADFQRYILY WRKRFTEQPI TDFCSVIRIN STAPFEEQEN YFLLCDVLPE DRILREELQK QRLREILEQQ QQERNDTNFH GVCMFCNEEF LGNRSVILNH MAREHAFNIG LPDNIVNCNE FLCTLQKKLD NLQCLYCEKT FRDKNTLKDH MRKKQHRKIN PKNREYDRFY VINYLVRLSF HNLKFDCSTA NKIFLRTN
ZNF277 (Zinc Finger Protein 277) is a Protein Coding gene. Diseases associated with ZNF277 includeSpecific Language ImpairmentandSpeech And Communication Disorders. Gene Ontology (GO) annotations related to this gene includeRNA polymerase II core promoter sequence specific DNA binding.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Nuclear receptor-interacting factor 4; zinc finger protein (C2H2 type) 277; Zinc finger protein 277; zinc finger protein 277 pseudogene
Gene Aliases: NRIF4; ZNF277; ZNF277P
UniProt ID: (Human) Q9NRM2
Entrez Gene ID: (Human) 11179
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support