Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MNSSSRSQIL LRWKSDKAQS GPYNVEKEIL TSRFLRDTET CRQNFRNFPY PDLAGPRKAL SQLRELCLKW LRPEIHSKEQ ILELLVLEQF LTILPGEVRT WVKSQYPESS EEAVTLVEDL TQILEEEAPQ NSTLSQDTPE EDPRGKHAFQ TGWLNDLVTK ESMTFKDVAV DITQEDWELM RPVQKELYKT VTLQNYWNMV SLGLTVYRPT VIPILEEPWM VIKEILEGPS PEWETKAQAC TPVEDMSKLT KEETHTIKLE DSYDYDDRLE RRGKGGFWKI HTDERGFSLK SVLSQEYDPT EECLSKYDIY RNNFEKHSNL IVQFDTQLDN KTSVYNEGRA TFNHVSYGIV HRKILPGEKP YKCNVCGKKF RKYPSLLKHQ STHAKEKSYE CEECGKEFRH ISSLIAHQRM HTGEKPYECH QCGKAFSQRA HLTIHQRIHT GEKPYKCDDC GKDFSQRAHL TIHQRTHTGE KPYKCLECGK TFSHSSSLIN HQRVHTGEKP YICNECGKTF SQSTHLLQHQ KIHTGKKPYK CNECWKVFSQ STYLIRHQRI HSGEKCYKCN ECGKAFAHSS TLIQHQTTHT GEKSYICNIC GKAFSQSANL TQHHRTHTGE KPYKCSVCGK AFSQSVHLTQ HQRIHNGEKP FKCNICGKAY RQGANLTQHQ RIHTGEKPYK CNECGKAFIY SSSLNQHQRT HTGERPYKCN ECDKDFSQRT CLIQHQRIHT GEKPYACRIC GKTFTQSTNL IQHQRVHTGA KHRN
This gene encodes a member of the krueppel family of zinc finger proteins, suggesting a role as a transcription factor. Its specific function has not been determined. This gene is located near the Smith-Magenis syndrome region on chromosome 17.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 287; Zinc finger protein with KRAB and SCAN domains 13
Gene Aliases: ZKSCAN13; ZNF287; ZSCAN45
UniProt ID: (Human) Q9HBT7
Entrez Gene ID: (Human) 57336
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support