Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: QPEPNKVNHI VTTKDDNVFS DSSEQVNFDS EDSSCLPEDL SLSKQLKIQV KEEPVEEAEE EAPEASTAPK EAGPSKEASL WPCEKCGKMF TVHKQLERHQ ELLCSVKPFI CHVCNKAFRT NFRLWSHFQS HMSQASEESA HKESEVCP
Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
ZNF295 contains 1 BTB (POZ) domain and 8 C2H2-type zinc fingers. ZNF295 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger and BTB domain-containing protein 21; Zinc finger protein 295
Gene Aliases: KIAA1227; ZBTB21; ZNF295
UniProt ID: (Human) Q9ULJ3
Entrez Gene ID: (Human) 49854
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support