Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: METQADLVSQ EPQALLDSAL LSKVPAFSDK DSLGDEMLAA ALLKAKSQEL VTFEDVAVYF IRKEWKRLEP AQRDLYRDVM LENYGNVFSL DRETRTENDQ EISEDTRSHG
ZNF3 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 8 C2H2-type zinc fingers and 1 KRAB domain. ZNF3 is involved in cell differentiation and/or proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C2-H2 type zinc finger protein; Zinc finger protein 3; Zinc finger protein HF.12; Zinc finger protein HZF3.1; Zinc finger protein KOX25
Gene Aliases: A8-51; HF.12; KOX25; PP838; Zfp113; ZNF3
UniProt ID: (Human) P17036
Entrez Gene ID: (Human) 7551
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support