Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide Sequence: MALCYGTFWG YPKMLEAANL MEGLVDIGPW VTLPRGQPEV LEWGLPKDQD SVAFEDVAVN FTHEEWALLG PSQKNLYRDV MRETIRNLNC IGMKWENQNI DDQ
ZNF44 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GIOT-2; gonadotropin inducible transcription repressor-2; Gonadotropin-inducible ovary transcription repressor 2; gonadotropin-inducible transcription repressor 2; Zinc finger protein 44; Zinc finger protein 55; Zinc finger protein 58; Zinc finger protein KOX7; zinc finger protein ZnFP12
Gene Aliases: GIOT-2; GIOT2; KOX7; ZNF; ZNF44; ZNF504; ZNF55; ZNF58
UniProt ID: (Human) P15621
Entrez Gene ID: (Human) 51710
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support