Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MQAVVPLNKM TAISPEPQTL ASTEQNEVPR VVTSGEQEAI LRGNAADAES FRQRFRWFCY SEVAGPRKAL SQLWELCNQW LRPDIHTKEQ ILELLVFEQF LTILPGEIRI WVKSQHPESS EEVVTLIEDL TQMLEEKDPV SQDSTVSQEE NSKEDKMVTV CPNTESCESI TLKDVAVNFS RGEWKKLEPF QKELYKEVLL ENLRNLEFLD FPVSKLELIS QLKWVELPWL LEEVSKSSRL GSVI
ZNF483 may be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 483; zinc finger protein HIT-10; Zinc finger protein with KRAB and SCAN domains 16
Gene Aliases: KIAA1962; ZKSCAN16; ZNF483; ZSCAN48
UniProt ID: (Human) Q8TF39
Entrez Gene ID: (Human) 158399
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support