Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAALRAPTQ QVFVAFEDVA IYFSQEEWEL LDEMQRLLYR DVMLENFAVM ASLGCWCGAV DEGTPSAESV SVEELSQGRT PKADTSTDKS HPCEICTPVL RDILQMIELQ ASPCGQKLYL GGAPRDFWMS SNLHQLQKLD NGEKLFKVDG DQASFMMNCR FHVSGKPFTF GEVGRDFSAT SGLLQHQVTP TIERPHSRIR HLRVPTGRKP LKYTESRKSF REKSVFIQHQ RADSGERPYK CSECGKSFSQ SSGFLRHRKA HGRTRTHECS ECGKSFSRKT HLTQHQRVHT GERPYDCSEC GKSFRQVSVL IQHQRVHTGE RPYECSECGK SFSHSTNLYR HRSAHTSTRP YECSECGKSF SHSTNLFRHW RVHTGVRPYE CSECGKAFSC NIYLIHHQRF HTGERPYVCS ECGKSFGQKS VLIQHQRVHT GERPYECSEC GKVFSQSSGL FRHRRAHTKT KPYECSECEK SFSCKTDLIR HQTVHTGERP YECSVCGKSF IRKTHLIRHQ TVHTNERPYE CDECGKSYSQ SSALLQHRRV HTGERPYECR ECGKSFTRKN HLIQHKTVHT GERPYECSEC GKSFSQSSGL LRHRRVHVQ
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 530
Gene Aliases: KIAA1508; ZNF530
UniProt ID: (Human) Q6P9A1
Entrez Gene ID: (Human) 348327
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support