Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEARSMLVPP QASVCFEDVA MAFTQEEWEQ LDLAQRTLYR EVTLETWEHI VSLGLFLSKS DVISQLEQEE DLCRAEQEAP RGTSGLSQVT GVVADGSFRF PVWDFT
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZNF544 is a zinc finger protein belonging to the Kruppel C2H2-type zinc-finger protein family. It localizes to the nucleus and may play a role in transcriptional regulation. ZNF544 is a 715 amino acid long protein that contains 13 C2H2-type zinc fingers and 1 KRAB domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 544
Gene Aliases: ZNF544
UniProt ID: (Human) Q6NX49
Entrez Gene ID: (Human) 27300
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support