Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MTEESEETVL YIEHRYVCSE CNQLYGSLEE VLMHQNSHVP QQHFELVGVA DPGVTVATDT ASGTGLYQTL VQESQYQCLE CGQLLMSPSQ LLEHQELHLK MMAPQEAVPA EPSPKAPPLS SSTIHYECVD CKALFASQEL WLNHRQTHLR ATPTKAPAPV VLGSPVVLGP PVGQARVAVE HSYRKAEEGG EGATVPSAAA TTTEVVTEVE LLLYKCSECS QLFQLPADFL EHQATHFPAP VPESQEPALQ QEVQASSPAE VPVSQPDPLP ASDHSYELRN GEAIGRDRRG RRARRNNSGE AGGAATQELF CSACDQLFLS PHQLQQHLRS HREGVFKCPL CSRVFPSPSS LDQHLGDHSS ESHFLCVDCG LAFGTEALLL AHRRAHTPNP LHSCPCGKTF VNLTKFLYHR RTHGVGGVPL PTTPVPPEEP VIGFPEPAPA ETGEPEAPEP PVSEETSAGP AAPGTYRCLL CSREFGKALQ LTRHQRFVHR LERRHKCSIC GKMFKKKSHV RNHLRTHTGE RPFPCPDCSK PFNSPANLAR HRLTHTGERP YRCGDCGKAF TQSSTLRQHR LVHAQHFPYR CQECGVRFHR PYRLLMHRYH HTGEYPYKCR ECPRSFLLRR LLEVHQLVVH AGRQPHRCPS CGAAFPSSLR LREHRCAAAA AQAPRRFECG TCGKKVGSAA RLQAHEAAHA AAGPGEVLAK EPPAPRAPRA TRAPVASPAA LGSTATASPA APARRRGLEC SECKKLFSTE TSLQVHRRIH TGERPYPCPD CGKAFRQSTH LKDHRRLHTG ERPFACEVCG KAFAISMRLA EHRRIHTGER PYSCPDCGKS YRSFSNLWKH RKTHQQQHQA AVRQQLAEAE AAVGLAVMET AVEALPLVEA IEIYPLAEAE GVQISG
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 574
Gene Aliases: FP972; ZNF574
UniProt ID: (Human) Q6ZN55
Entrez Gene ID: (Human) 64763
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support