Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MKAKRRRPDL DEIHRELRPQ GSARPQPDPN AEFDPDLPGG GLHRCLACAR YFIDSTNLKT HFRSKDHKKR LKQLSVEPYS QEEAERAAGM GSYVPPRRLA VPTEVSTEVP EMDTST
Negatively modulates the DNA binding activity of Oct-2 and therefore its transcriptional regulatory activity. Could act either by binding to DNA octamer or by interacting with Oct-2. May also be a modulator of other octamer-binding proteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc Finger 593; Zinc finger protein 593; Zinc finger protein T86
Gene Aliases: ZNF593; ZT86
UniProt ID: (Human) O00488
Entrez Gene ID: (Human) 51042
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support