Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Sequence of this protein is as follows: FTRKHGNLTV DHIQNNSDSG AKKPTPPDYY PECQSQPAVS SPHHIIPQKQ HYAPPPSPSS PVNHQMPYPP QDVVTPNSVR SQVPALTTTY DPSSGYIIVK VPPDMNSPPL RAPQSQNGNP
ZNF645 contains 1 RING-type zinc finger, which is probably involved in mediating protein-protein interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: c-Cbl-like protein 2; Cbl proto-oncogene-like protein 2; E3 ubiquitin ligase; E3 ubiquitin-protein ligase CBLL2; RING-type E3 ubiquitin transferase ZNF645; Zinc finger protein 645
Gene Aliases: CBLL2; CT138; HAKAIL; ZNF645
UniProt ID: (Human) Q8N7E2
Entrez Gene ID: (Human) 158506
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support