Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEVVTFGDVA VHFSREEWQC LDPGQRALYR EVMLENHSSV AGLAGFLVFK PELISRLEQG EEPWVLDLQG AEGTEAPRTS KTDSTIRTEN EQACEDMDIL KSESYGTVVR ISPQDFPQNP GFGDVSDSEV WLDSHLGSPG LKVTGFTFQN NCLNEETVVP KTFTKDAPQG CKELGSSGLD CQPLESQGES AEGMSQRCEE CGKGIRATSD IALHWEINTQ KISRCQECQK KLSDCLQGKH TNNCHGEKPY ECAECGKVFR LCSQLNQHQR IHTGEKPFKC TECGKAFRLS SKLIQHQRIH TGEKPYRCEE CGKAFGQSSS LIHHQRIHTG ERPYGCRECG KAFSQQSQLV RHQRTHTGER PYPCKECGKA FSQSSTLAQH QRMHTGEKAQ ILKASDSPSL VAHQRIHAVE KPFKCDECGK AFRWISRLSQ HQLIHTGEKP YKCNKCTKAF GCSSRLIRHQ RTHTGEKPFK CDECGKGFVQ GSHLIQHQRI HTGEKPYVCN DCGKAFSQSS SLIYHQRIHK GEKPYECLQC GKAFSMSTQL TIHQRVHTGE RPYKCNECGK AFSQNSTLFQ HQIIHAGVKP YECSECGKAF SRSSYLIEHQ RIHTRAQWFY EYGNALEGST FVSRKKVNTI KKLHQCEDCE KIFRWRSHLI IHQRIHTGEK PYKCNDCGKA FNRSSRLTQH QKIHMG
May be involved in transcriptional regulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C2-H2 type zinc finger protein; Zinc finger protein 7; zinc finger protein 7 (KOX 4, clone HF.16); Zinc finger protein HF.16; Zinc finger protein KOX4; zinc finger protein zfp30; Zinc finger protein-7 (KOX4)
Gene Aliases: HF.16; KOX4; zf30; ZNF7
UniProt ID: (Human) B4DT08
Entrez Gene ID: (Human) 7553
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support