Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEEEEMNDGS QMVRSQESLT FQDVAVDFTR EEWDQLYPAQ KNLYRDVMLE NYRNLVALGY QLCKPEVIAQ LELEEEWVIE RDSLLDTHPD GENRPEIKKS TTSQNISDEN QTHEMIMERL AGDSFWYSIL GGLWDFDYHP EFNQENHKRY LGQVTLTHKK ITQERSLECN KFAENCNLNS NLMQQRIPSI KIPLNSDTQG NSIKHNSDLI YYQGNYVRET PYEYSECGKI FNQHILLTDH IHTAEKPSEC GKAFSHTSSL SQPQMLLTGE KPYKCDECGK RFSQRIHLIQ HQRIHTGEKP FICNGCGKAF RQHSSFTQHL RIHTGEKPYK CNQCGKAFSR ITSLTEHHRL HTGEKPYECG FCGKAFSQRT HLNQHERTHT GEKPYKCNEC GKAFSQSAHL NQHRKIHTRE KLCEYKCEQT VRHSPSFSST
ZNF713 is a new candidate transcription factor.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Zinc finger protein 713
Gene Aliases: ZNF713
UniProt ID: (Human) Q8N859
Entrez Gene ID: (Human) 349075
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support