Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide Sequence: MASTWAIQAH MDQDEPLEVK IEEEKYTTRQ DWDLRKNNTH SREVFRQYFR QFCYQETSGP REALSRLREL CHQWLRPETH TKEQILELLV LEQFLTILPE ELQAWVQEQH PESGEEVVTV LEDLERELDE PGEQVSVHTG EQEMFLQETV RLRKEGEPSM SLQSMKAQPK YESPELESQQ EQVLDVETGN EYGNLKQEVS EEMEPHGKTS SKFENDMSKS ARCGETREPE EITEEPSACS REDKQPTCDE NGVSLTENSD HTEHQRICPG EESYGCDDCG KAFSQHSHLI EHQRIHTGDR PYKCEECGKA FRGRTVLIRH KIIHTGEKPY KCNECGKAFG RWSALNQHQR LHTGEKHYHC NDCGKAFSQK AGLFHHIKIH TRDKPYQCTQ CNKSFSRRSI LTQHQGVHTG AKPYECNECG KAFVYNSSLV SHQEIHHKEK CYQCKECGKS FSQSGLIQHQ RIHTGEKPYK CDVCEKAFIQ RTSLTEHQRI HTGERPYKCD KCGKAFTQRS VLTEHQRIEC GNAFRGITSL IQHQRIHTGE KPYQCDECGK AFRQRKKTSY KEILLKNHSE PQAGVNLLLS SLIPEWQSCF RRDL
May be involved in transcriptional regulation. Classified as a zinc finger protein expressed in the corpus luteum.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DM9; Zinc finger and SCAN domain-containing protein 12; Zinc finger protein 305; Zinc finger protein 96
Gene Aliases: dJ29K1.2; KIAA0426; ZFP96; ZNF29K1; ZNF305; ZNF96; ZSCAN12
UniProt ID: (Human) O43309
Entrez Gene ID: (Human) 9753
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support