Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MASLKCSTVV CVICLEKPKY RCPACRVPYC SVVCFRKHKE QCNPETRPVE KKIRSALPTK TVKPVENKDD DDSIADFLNS DEEEDRVSLQ NLKNLGESAT LRSLLLNPHL RQLMVNLDQG EDKAKLMRAY MQEPLFVEFA DCCLGIVEPS QNEES
ZNHIT3 contains 1 HIT-type zinc finger. The exact function of ZNHIT3 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HNF-4a coactivator; Thyroid hormone receptor interactor 3; thyroid receptor interacting protein 3; Thyroid receptor-interacting protein 3; TR-interacting protein 3; TRIP-3; Zinc finger HIT domain-containing protein 3; zinc finger, HIT domain containing 3; zinc finger, HIT type 3; zinc finger, HIT-type containing 3
Gene Aliases: TRIP3; ZNHIT3
UniProt ID: (Human) Q15649
Entrez Gene ID: (Human) 9326
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support