Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSVMDLANTC SSFQSDLDFC SDCGSVLPLP GAQDTVTCIR CGFNINVRDF EGKVVKTSVV FHQLGTAMPM SVEEGPECQG PVVDRRCPRC GHEGMAYHTR QMRSADEGQT VFYTCTNCKF QEKEDS
This gene encodes a DNA-directed RNA polymerase I subunit. The encoded protein contains two potential zinc-binding motifs and may play a role in regulation of cell proliferation. The encoded protein may be involved in cancer and human immunodeficiency virus progression. Alternative splicing results in multiple transcript variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-directed RNA polymerase I subunit H; DNA-directed RNA polymerase I subunit RPA12; RNA polymerase I small specific subunit Rpa12; transcription-associated zinc ribbon protein; Zinc ribbon domain-containing protein 1
Gene Aliases: HTEX-6; HTEX6; hZR14; POLR1H; RPA12; tctex-6; TCTEX6; TEX6; ZNRD1; ZR14
UniProt ID: (Human) Q9P1U0
Entrez Gene ID: (Human) 30834
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support