Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGGKQSTAAR SRGPFPGVST DDSAVPPPGG APHFGHYRTG GGAMGLRSRS VSSVAGMGMD PSTAGGVPFG LYTPASRGTG DSERAPGGGG SASDSTYAHG NGYQETGGGH HRDGMLYLGS RASLADALPL HIAPRWFSSH SGFKCPICSK SVASDEMEMH FIMCLSKPRL SYNDDVLTKD AGECVICLEE LLQGDTIARL PCLCIYHKSC IDSWFEVNRS CPEHPAD
E3 ubiquitin-protein ligase that mediates the ubiquitination of AKT1 and GLUL, thereby playing a role in neuron cells differentiation. Plays a role in the establishment and maintenance of neuronal transmission and plasticity. Regulates Schwann cells differentiation by mediating ubiquitination of GLUL. Promotes neurodegeneration by mediating 'Lys-48'-linked polyubiquitination and subsequent degradation of AKT1 in axons: degradation of AKT1 prevents AKT1-mediated phosphorylation of GSK3B, leading to GSK3B activation and phosphorylation of DPYSL2/CRMP2 followed by destabilization of microtubule assembly in axons (Probable).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase ZNRF1; MGC101991; nerve injury gene 283; Nerve injury-induced gene 283 protein; RING-type E3 ubiquitin transferase ZNRF1; zinc and ring finger protein 1; Zinc/RING finger protein 1
Gene Aliases: NIN283; ZNRF1
UniProt ID: (Human) Q8ND25
Entrez Gene ID: (Human) 84937
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support