Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bethyl Laboratories
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
Human DNA-binding protein (dbpA) is a member of a Y-box binding protein family containing a cold shock domain. The increased expression of Y box binding proteins in somatic cells is associated with cell proliferation and transformation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cold shock domain protein A; Cold shock domain-containing protein A; cold-shock domain containing A1; cold-shock domain protein A; DNA-binding protein A; Single-strand DNA-binding protein NF-GMB; Y box binding protein 3; Y-box-binding protein 3; ZO-1-associated nucleic acid-binding protein; ZONAB A; ZONAB B
Gene Aliases: CSDA; CSDA1; DBPA; YBX3; ZONAB
UniProt ID: (Human) P16989
Entrez Gene ID: (Human) 8531
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support