Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
OriGene
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
ZSCAN21 belongs to the krueppel C2H2-type zinc-finger protein family and is a strong transcriptional activator.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CtFIN51; Renal carcinoma antigen NY-REN-21; Transcription factor RU49; Zfp-38; Zinc finger and SCAN domain-containing protein 21; zinc finger proliferation 1; Zinc finger protein 38; zinc finger protein 38 (KOX 25); Zinc finger protein 38 homolog; zinc finger protein NY-REN-21 antigen
Gene Aliases: AI326272; CTfin51; NY-REN-21; RU49; Zfp-38; ZFP38; Zipro1; ZNF38; ZSCAN21
UniProt ID: (Human) Q9Y5A6, (Mouse) Q07231
Entrez Gene ID: (Human) 7589, (Mouse) 22697
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support