Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LADVLEETHY ENGEYIIRQG ARGDTFFIIS KGTVNVTRED SPSEDPVFLR TLGKGDWFGE KALQGEDVRT ANVIAAEAVT CLVIDRDSFK HLIGGLDDVS NKAYEDAEAK AKYEAEA
PRKG1 is a cGMP-dependent serine/threonine kinase which has an important role in the relaxation of vascular smooth muscle and the inhibition of platelet aggregation. The PRKG1 proteins play a central role in regulating cardiovascular and neuronal functions in addition to relaxing smooth muscle tone, and modulating cell growth. This gene is most strongly expressed in all types of smooth muscle, platelets, cerebellar Purkinje cells, hippocampal neurons, and the lateral amygdala. Isoforms Ialpha and Ibeta have identical cGMP-binding and catalytic domains but differ in their leucine/isoleucine zipper and autoinhibitory sequences and therefore differ in their dimerization substrates and kinase enzyme activity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cGK 1; cGK cGK cGKI; cGK cGK1; cGKI; cGMP-dependent protein kinase 1; cGMP-dependent protein kinase I; OTTHUMP00000019618; protein kinase, cGMP-dependent, regulatory, type I, beta; RP11-346D6.1
Gene Aliases: AAT8; cGK; cGK 1; cGK1; cGKI; cGKI-alpha; cGKI-BETA; PKG; PRKG1; PRKG1B; PRKGR1A; PRKGR1B
UniProt ID: (Human) Q13976
Entrez Gene ID: (Human) 5592
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support