Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • cGKI Antibodies

          Invitrogen

          cGKI Polyclonal Antibody

          View all (21) cGKI antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite cGKI Polyclonal Antibody

          • Antibody Testing Data (1)
          cGKI Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          cGKI Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          cGKI Antibody (PA5-96164) in WB

          Western blot analysis of cGKI in extracts of HepG2 cells. Samples were incubated with cGKI Polyclonal antibody (Product # PA5-96164) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) at a dilution of 1:10,000. Lysates/proteins: 25 µg per lane. Blocking buffer: 3% nonfat dry milk in TBST. Detection: ECL Basic Kit. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          cGKI Antibody in Western Blot (WB)

          Product Details

          PA5-96164

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human PRKG1 (NP_0062491)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.85 mg/mL

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          RRID

          AB_2807966

          Product Specific Information

          Immunogen sequence: MGTLRDLQYA LQEKIEELRQ RDALIDELEL ELDQKDELIQ KLQNELDKYR SVIRPATQQA QKQSASTLQG EPRTKRQAIS AEPTAFDIQD LSHVTLPFYP KSPQSKDLIK EAILDNDFMK NLELSQIQEI VDCMYPVEYG KDSCIIKEGD VGSLVYVMED GKVEVTKEGV KLCTMGPGKV FGELAILYNC TRTATVKTLV NVKLWAIDRQ CFQTIMMRTG LIKHTEYMEF LKSVPTFQSL PEEILSKLAD VLEETHYENG EYIIRQGARG DTFFIISKGT VNVTREDSPS EDPVFLRTLG; Positive Samples: HepG2; Cellular Location: Cytoplasm

          Target Information

          PRKG1 is a cGMP-dependent serine/threonine kinase which has an important role in the relaxation of vascular smooth muscle and the inhibition of platelet aggregation. The PRKG1 proteins play a central role in regulating cardiovascular and neuronal functions in addition to relaxing smooth muscle tone, and modulating cell growth. This gene is most strongly expressed in all types of smooth muscle, platelets, cerebellar Purkinje cells, hippocampal neurons, and the lateral amygdala. Isoforms Ialpha and Ibeta have identical cGMP-binding and catalytic domains but differ in their leucine/isoleucine zipper and autoinhibitory sequences and therefore differ in their dimerization substrates and kinase enzyme activity.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: cGK 1; cGK cGK cGKI; cGK cGK1; cGK1; cGKI; cGMP-dependent protein kinase 1; cGMP-dependent protein kinase I; OTTHUMP00000019618; protein kinase, cGMP-dependent, regulatory, type I, beta; RP11-346D6.1

          View more View less

          Gene Aliases: AAT8; AW125416; cGK; cGK 1; cGK1; CGKI; cGKI-alpha; cGKI-BETA; Gm19690; PKG; PRKG1; PRKG1B; PRKGR1A; PRKGR1B

          View more View less

          UniProt ID: (Human) Q13976, (Mouse) P0C605

          View more View less

          Entrez Gene ID: (Human) 5592, (Mouse) 19091

          View more View less

          Function(s)
          protein serine/threonine kinase activity cGMP-dependent protein kinase activity calcium channel regulator activity protein binding ATP binding cGMP binding nucleotide binding catalytic activity protein kinase activity kinase activity transferase activity transferase activity, transferring phosphorus-containing groups protein homodimerization activity mitogen-activated protein kinase p38 binding
          Process(es)
          neuron migration protein phosphorylation signal transduction positive regulation of cytosolic calcium ion concentration metabolic process negative regulation of inositol phosphate biosynthetic process negative regulation of glutamate secretion negative regulation of smooth muscle cell migration phosphorylation dendrite development cGMP-mediated signaling forebrain development positive regulation of circadian rhythm regulation of GTPase activity negative regulation of heart contraction positive regulation of vasodilation negative regulation of smooth muscle contraction relaxation of vascular smooth muscle cell growth involved in cardiac muscle cell development negative regulation of platelet aggregation mitophagy in response to mitochondrial depolarization positive regulation of large conductance calcium-activated potassium channel activity regulation of testosterone biosynthetic process actin cytoskeleton organization
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-k2m5x:80/100.66.128.182:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0