Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen Sequence: LRKKVWINTS DIILVGLRDY QDNKADVILK YNADEARSLK AYGELPEHAK INETDTFGPG DDDEIQFDDI GDDDEDIDDI
This gene encodes an essential eukaryotic translation initiation factor. The protein is required for the binding of the 43S complex (a 40S subunit, eIF2/GTP/Met-tRNAi and eIF3) to the 5' end of capped RNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-1A X isoform; eIF-4C; eukaryotic translation initiation factor 1A, X chromosome; Eukaryotic translation initiation factor 1A, X-chromosomal; eukaryotic translation initiation factor 1A, Y-linked; Eukaryotic translation initiation factor 4C; Putative eukaryotic translation initiation factor 1A
Gene Aliases: 1500010B24Rik; AI426898; eIF-1A; eIF-4C; EIF1A; EIF1AP1; EIF1AX; Eif1ay; EIF4C
UniProt ID: (Human) P47813, (Mouse) Q8BMJ3
Entrez Gene ID: (Human) 1964, (Mouse) 66235
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support