Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Positive Samples: U-251MG
Immunogen sequence: VDGRVVEPKR AVSREDSVKP GAHLTVKKIF VGGIKEDTEE YNLRDYFEKY GKIETIEVME DRQSGKKRGF AFVTFDDHDT VDKIVVQKYH TINGHNCEVK K
HNRPA3 contains 2 RRM (RNA recognition motif) domains and plays a role in cytoplasmic trafficking of RNA. It binds to the cis-acting response element, A2RE and may be involved in pre-mRNA splicing.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heterogeneous nuclear ribonucleoprotein A3; hnRNP A3
Gene Aliases: 2410013L13Rik; 2610209F03Rik; 2610510D13Rik; D10S102; FBRNP; HNRNPA3; HNRPA3
UniProt ID: (Human) P51991, (Mouse) Q8BG05, (Rat) Q6URK4
Entrez Gene ID: (Human) 220988, (Mouse) 229279, (Rat) 362152
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support