Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive test controls include: Rat testis, Rat brain. The target is usually found in the following locations: Nucleus, nucleoplasm.
Immunogen sequence: VRTHYDPPRK LMAMQRPGPY DRPGAGRGYN SIGRGAGFER MRRGAYGGGY GGYDDYNGYN DGYGFGSDRF GRDLNYCFSG MSDHRYGDGG STFQSTTGHC VHMRGLPYRA TENDIYNFFS PLNPVRVHIE IGPDGRVTGE ADVEFATHED AVAAMSKDKA NMQHRYVELF LNSTAGASGG AYEHRYVELF LNSTAGASGG AYGSQMMGGM GLSNQSSYGG PASQQLSGGY GGGYGGQSSM SGYDQVLQEN SSDFQSNIA
This gene encodes a member of a subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins that complex with heterogeneous nuclear RNA. These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some may shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that bind to RNA and is very similar to the family member HNRPF. This gene may be associated with hereditary lymphedema type I. Alternatively spliced transcript variants have been described.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Heterogeneous nuclear ribonucleoprotein H; hnRNP H; Ratsg1
Gene Aliases: AI642080; E430005G16Rik; Hnrnph; HNRNPH1; HNRPH; HNRPH1
UniProt ID: (Human) P31943, (Mouse) O35737, (Rat) Q8VHV7
Entrez Gene ID: (Human) 3187, (Mouse) 59013, (Rat) 140931
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support