Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • hnRNP L Antibodies

          Invitrogen

          hnRNP L Polyclonal Antibody

          View all (26) hnRNP L antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite hnRNP L Polyclonal Antibody

          • Antibody Testing Data (2)
          hnRNP L Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          hnRNP L Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          hnRNP L Antibody (PA5-97050) in ICC/IF

          Immunofluorescence analysis of hnRNP L in 293T cells. Samples were incubated with hnRNP L Polyclonal antibody (Product # PA5-97050) using a dilution of 1:100 (40x lens). Blue: DAPI for nuclear staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          hnRNP L Antibody in Immunocytochemistry (ICC/IF)
          hnRNP L Antibody in Western Blot (WB)

          Product Details

          PA5-97050

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:100
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant fusion protein containing a sequence corresponding to amino acids 282-589 of human HNRNPL (NP_0015242)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          2.93 mg/mL

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          RRID

          AB_2808852

          Product Specific Information

          Immunogen sequence: TWDYTNPNLS GQGDPGSNPN KRQRQPPLLG DHPAEYGGPH GGYHSHYHDE GYGPPPPHYE GRRMGPPVGG HRRGPSRYGP QYGHPPPPPP PPEYGPHADS PVLMVYGLDQ SKMNCDRVFN VFCLYGNVEK VKFMKSKPGA AMVEMADGYA VDRAITHLNN NFMFGQKLNV CVSKQPAIMP GQSYGLEDGS CSYKDFSESR NNRFSTPEQA AKNRIQHPSN VLHFFNAPLE VTEENFFEIC DELGVKRPSS VKVFSGKSER SSSGLLEWES KSDALETLGF LNHYQMKNPN GPYPYTLKLC FSTAQHAS; Positive Samples: U-87MG, HeLa, HepG2, A-549, HT-29, Mouse ovary, Rat lung, Rat brain; Cellular Location: Cytoplasm, Nucleus, nucleoplasm

          Target Information

          Heterogeneous nuclear RNAs (hnRNAs) which include mRNA precursors and mature mRNAs are associated with specific proteins to form heterogenous ribonucleoprotein (hnRNP) complexes. Heterogeneous nuclear ribonucleoprotein L is among the proteins that are stably associated with hnRNP complexes and along with other hnRNP proteins is likely to play a major role in the formation, packaging, processing, and function of mRNA. Heterogeneous nuclear ribonucleoprotein L is present in the nucleoplasm as part of the HNRP complex. HNRP proteins have also been identified outside of the nucleoplasm. Exchange of hnRNP for mRNA-binding proteins accompanies transport of mRNA from the nucleus to the cytoplasm. Since HNRP proteins have been shown to shuttle between the nucleus and the cytoplasm, it is possible that they also have cytoplasmic functions. Two transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: FLJ35509; Heterogeneous nuclear ribonucleoprotein L; heterogenous nuclear ribonucleoprotein L; hnRNP L

          View more View less

          Gene Aliases: C79783; D830027H13Rik; Fblim1; hnRNP-L; HNRNPL; HNRPL; P/OKcl.14

          View more View less

          UniProt ID: (Human) P14866, (Mouse) Q8R081, (Rat) F1LQ48

          View more View less

          Entrez Gene ID: (Human) 3191, (Mouse) 15388, (Rat) 80846

          View more View less

          Function(s)
          nucleotide binding RNA binding protein binding transcription regulatory region DNA binding poly(A) RNA binding pre-mRNA intronic binding nucleic acid binding mRNA 3'-UTR binding
          Process(es)
          mRNA processing cellular response to amino acid starvation positive regulation of translation negative regulation of mRNA splicing, via spliceosome positive regulation of mRNA binding regulation of alternative mRNA splicing, via spliceosome mRNA splicing, via spliceosome RNA processing gene expression
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67c77f6dc7-n6nf2:80/100.66.135.186:80.
          git-commit: 545e792480003146e539d624bd2a21b51d198228
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0