Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • mGluR1/mGluR5 Antibodies

          QED Bioscience

          mGluR1/5 Glutamate Receptor Monoclonal Antibody (S75-3)

          View all (12) mGluR1/mGluR5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite mGluR1/5 Glutamate Receptor Monoclonal Antibody (S75-3)

          • Antibody Testing Data (3)
          mGluR1/5 Glutamate Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          mGluR1/5 Glutamate Receptor Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          mGluR1/5 Glutamate Receptor Antibody (56545-100UG) in ICC/IF

          Immunocytochemistry/Immunofluorescence analysis using Mouse Anti-mGluR1/5 glutamate receptor Monoclonal Antibody, Clone S75-3 (56545). Tissue: Neuroblastoma cell line (SK-N-BE). Species: Human. Fixation: 4% Formaldehyde for 15 min at RT. Primary Antibody: Mouse Anti-mGluR1/5 glutamate receptor Monoclonal Antibody (56545) at 1:100 for 60 min at RT. Secondary Antibody: Goat Anti-Mouse ATTO 488 at 1:200 for 60... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          mGluR1/5 Glutamate Receptor Antibody in Immunocytochemistry (ICC/IF)
          mGluR1/5 Glutamate Receptor Antibody in Immunocytochemistry (ICC/IF)
          mGluR1/5 Glutamate Receptor Antibody in Western Blot (WB)

          Product Details

          56545-100UG

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          1:50
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          S75-3

          Immunogen

          Fusion protein corresponding to aa 824-1203 (cytoplasmic C-terminus) of rat mGluR5b (accession no. P31424).
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1.0 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Target Information

          may play a role in synaptic transmission; may interact with mGluR1, another group I metabotropic glutamate receptor.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: G protein coupled receptor family C group 1 member A; G protein coupled receptor, family C, group 1, member A; G protein coupled receptor, family C, group 1, member E; G protein-coupled receptor GRM5; G protein-coupled receptor, family C, group 1, member A; G protein-coupled receptor, family C, group 1, member E; glutamate receptor; glutamate receptor, metabotropic 1; glutamate receptor, metabotropic 5; metabotropic glutamate receptor (mGluR5); Metabotropic glutamate receptor 1; Metabotropic glutamate receptor 5; metabotropic glutamate receptor 5b; metabotropic glutamate receptor subtype 5b; metabotropic glutamate receptor type 5; mGluR1; mGluR5; mGlurR1a; protein phosphatase 1, regulatory subunit 85; protein phosphatase 1, regulatory subunit 86

          View more View less

          Gene Aliases: 4930455H15Rik; 6430542K11Rik; AI850523; ENSMUSG00000075319; Glu5R; Gm10828; GPRC1A; GPRC1E; GRM1; GRM5; MGLU1; mGlu5; MGLUR1; MGLUR5; mGluR5b; nmf373; PPP1R85; PPP1R86; rcw; SCAR13; wobl

          View more View less

          UniProt ID: (Human) Q13255, (Human) P41594, (Mouse) P97772, (Rat) P23385, (Rat) P31424, (Mouse) Q3UVX5

          View more View less

          Entrez Gene ID: (Human) 2911, (Human) 2915, (Mouse) 14816, (Rat) 24414, (Rat) 24418, (Mouse) 108071

          View more View less

          Function(s)
          G-protein coupled receptor activity protein binding glutamate receptor activity PLC activating G-protein coupled glutamate receptor activity signal transducer activity estrogen receptor binding identical protein binding calmodulin binding PDZ domain binding A2A adenosine receptor binding
          Process(es)
          activation of MAPKKK activity desensitization of G-protein coupled receptor protein signaling pathway regulation of transcription, DNA-templated protein phosphorylation protein kinase C-activating G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled glutamate receptor signaling pathway G-protein coupled glutamate receptor signaling pathway chemical synaptic transmission learning locomotory behavior negative regulation of locomotion phosphatidylinositol-mediated signaling regulation of long-term neuronal synaptic plasticity positive regulation of long-term neuronal synaptic plasticity cognition positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway regulation of synaptic transmission, glutamatergic signal transduction G-protein coupled receptor signaling pathway activation of MAPKK activity activation of MAPK activity sensory perception of pain calcium-mediated signaling dimeric G-protein coupled receptor signaling pathway regulation of MAPK cascade regulation of sensory perception of pain cellular response to electrical stimulus regulation of glutamate secretion membrane depolarization synaptic transmission, GABAergic
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0