Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • p14ARF Antibodies

          NeoBiotechnologies

          p14ARF Monoclonal Antibody (4C6/4)

          View all (30) p14ARF antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite p14ARF Monoclonal Antibody (4C6/4)

          • Antibody Testing Data (3)
          p14ARF Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          p14ARF Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          p14ARF Antibody (1029-MSM1-P0) in IHC (P)

          Formalin-fixed, paraffin-embedded human Brain stained with p14ARF Mouse Monoclonal Antibody (4C6/4). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          p14ARF Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          p14ARF Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          p14ARF Antibody in SDS-PAGE (SDS-PAGE)

          Product Details

          1029-MSM1-P0

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-2 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          Assay-dependent
          -

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          1-2 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          1-2 µg/mL
          -

          Flow Cytometry (Flow)

          1-2 µg/10^6 cells
          -
          Product Specifications

          Species Reactivity

          Human, Rat

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          4C6/4

          Immunogen

          Recombinant full-length human p14 ARF protein
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          200 µg/mL

          Purification

          Protein A/G

          Storage buffer

          PBS, pH 7.4, with 0.05% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The INK4a locus on chromosome 9p21 is frequently affected in human tumors. It encodes two structurally distinct tumor suppressor proteins, p16INK4a and the alternative reading frame protein, ARF (p14ARF in human and P19ARF in mouse). These two proteins interact with the upstream proteins: retinoblastoma protein and p53 tumor suppressors, respectively. Each of these proteins has a role in the senescence of primary cells, activates pathways for cell cycle control and tumor suppression. The p14ARF (or P19ARF in mouse) proteins can induce both G1 and G2 phase arrest in a manner that depends on functional p53. The mouse and human ARF proteins have different functions in tumor suppression; this distinction may contribute to the different levels of tumor proneness of these species.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Alternative reading frame; ARF; CDK4 inhibitor p16-INK4; CDK4I; cdkn2a {ECO:0000312|EMBL:AAL76336.1, ECO:0000312|RGD:2323}; cdkn2a {ECO:0000312|EMBL:AAL76338.1, ECO:0000312|RGD:2323}; cell cycle inhibitor; cell cycle negative regulator beta; cell cycle regulator; Cyclin dependent kinase inhibitor 2A (p16, inhibits CDK4); Cyclin-dependent kinase 4 inhibitor A; Cyclin-dependent kinase inhibitor 2A; cyclin-dependent kinase inhibitor 2A (melanoma, p16, inhibits CDK4); cyclin-dependent kinase inhibitor 2a p16Ink4a; cyclin-dependent kinase inhibitor 2a p19Arf; cyclin-dependent kinase inhibitor 2A, isoform 1; cyclin-dependent kinase inhibitor 2A, isoform 2; inhibits CDK4; MTS-1; Multiple tumor suppressor 1; p14 p16; p14ARF; p16; p16-INK4; p16-INK4a; P19 TP16; Tumor suppressor ARF

          View more View less

          Gene Aliases: ARF; CDK4I; CDKN2; CDKN2A; CMM2; INK4; INK4A; MLM; MTS-1; MTS1; P14; P14ARF; P16; P16-INK4A; p16Cdkn2a; P16INK4; P16INK4A; P19; P19ARF; TP16

          View more View less

          UniProt ID: (Human) P42771, (Rat) Q9R0Z3

          View more View less

          Entrez Gene ID: (Human) 1029, (Rat) 25163

          View more View less

          Function(s)
          cyclin-dependent protein serine/threonine kinase inhibitor activity protein binding protein kinase binding poly(A) RNA binding NF-kappaB binding p53 binding DNA binding transcription factor activity, sequence-specific DNA binding transcription factor binding ubiquitin-protein transferase inhibitor activity MDM2/MDM4 family protein binding
          Process(es)
          regulation of cyclin-dependent protein serine/threonine kinase activity G1/S transition of mitotic cell cycle protein polyubiquitination negative regulation of cell-matrix adhesion transcription, DNA-templated regulation of transcription, DNA-templated rRNA processing negative regulation of protein kinase activity apoptotic process activation of cysteine-type endopeptidase activity involved in apoptotic process cell cycle arrest Ras protein signal transduction aging cell aging negative regulation of cell proliferation epidermis development apoptotic mitochondrial changes rRNA transcription response to organic substance response to organonitrogen compound regulation of G2/M transition of mitotic cell cycle regulation of gene expression response to organic cyclic compound cerebellum development negative regulation of cell growth negative regulation of B cell proliferation regulation of protein stability protein destabilization negative regulation of NF-kappaB transcription factor activity negative regulation of protein binding negative regulation of immature T cell proliferation in thymus positive regulation of protein sumoylation negative regulation of mammary gland epithelial cell proliferation positive regulation of smooth muscle cell apoptotic process somatic stem cell population maintenance senescence-associated heterochromatin focus assembly negative regulation of phosphorylation response to drug glucose homeostasis positive regulation of apoptotic process positive regulation of DNA damage response, signal transduction by p53 class mediator negative regulation of cyclin-dependent protein serine/threonine kinase activity negative regulation of cell cycle negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter regulation of nucleocytoplasmic transport regulation of protein export from nucleus somatic stem cell division protein stabilization positive regulation of sequence-specific DNA binding transcription factor activity negative regulation of ubiquitin-protein transferase activity positive regulation of apoptotic process involved in mammary gland involution cellular response to hydrogen peroxide protein K63-linked ubiquitination positive regulation of cell cycle arrest cellular senescence replicative senescence positive regulation of protein localization to nucleus positive regulation of signal transduction by p53 class mediator regulation of apoptotic DNA fragmentation negative regulation of proteolysis involved in cellular protein catabolic process negative regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process positive regulation of macrophage apoptotic process positive regulation of cellular senescence
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0